Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
METTL7A Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177336PCP
Description
METTL7A Polyclonal antibody specifically detects METTL7A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
METTL7A | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
AAM-B, DKFZP586A0522, EC 2.1.1, EC 2.1.1.-, methyltransferase like 7A, methyltransferase-like protein 7A, Protein AAM-B | |
Antibody was raised against a 13 amino acid synthetic peptide near the center of human MettL7A. The immunogen is located within amino acids 130 - 180 of MettL7A. Amino Acid Squence: SVKNQERILREV | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA, Immunocytochemistry | |
PerCP | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
25840 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction