Learn More
METTL7A Monoclonal Antibody (OTI4B10)

Description
METTL7A Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | METTL7A |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI4B10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | METTL7A |
| Gene Accession No. | Q9H8H3 |
| Gene Alias | 2210414H16Rik; 3300001H21Rik; Aam-B; methyltransferase like 7A; methyltransferase like 7A1; methyltransferase like 7A-like; methyltransferase-like protein 7A; METTL7A; Mettl7a1; PRO0066; Protein AAM-B; RGD1308407; UbiE1; UNQ1902/PRO4348 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.