Learn More
Invitrogen™ METTL7A Polyclonal Antibody

Description
A suggested positive control is MCF7 cell lysate. PA5-20636 can be used with blocking peptide PEP-0756.
Specifications
Specifications
| Antigen | METTL7A |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | METTL7A |
| Gene Accession No. | Q9H8H3 |
| Gene Alias | 2210414H16Rik; 3300001H21Rik; Aam-B; methyltransferase like 7A; methyltransferase like 7A1; methyltransferase like 7A-like; methyltransferase-like protein 7A; METTL7A; Mettl7a1; PRO0066; Protein AAM-B; RGD1308407; UbiE1; UNQ1902/PRO4348 |
| Gene Symbols | METTL7A, Mettl7a1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.