Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METTL7A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596971
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596971 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596971 Supplier Invitrogen™ Supplier No. PA596971
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WICKKWFPYF LVRFTVIYNE QMASKKRELF SNLQEFAGPS GKLSLLEVGC GTGANFKFYP PGCRVTCIDP NPNFEKFLIK SIAENRHLQF ERFVVAAGEN MHQVADGSVD VVVCTLVLCS VKNQERILRE VCRVLRPGGA FYFMEHVAAE CSTWNYFWQQ VLDPAWHLLF DGCNLTRESW KALERASFSK LKLQHIQAPL SWELVRPHIY GYAVK; Positive Samples: 22Rv1; Cellular Location: Endoplasmic reticulum, Lipid droplet, Membrane.

MettL7A belongs to the methyltransferase superfamily. It is a probable methyltransferase. Methyltransferase is a type of transferase enzyme which transfers a methyl group from a donor to an acceptor. Often methylation occurs on nucleic bases in DNA or amino acids in protein structures. DNA methylation is often utilized to silence and regulate genes without changing the original DNA sequence. DNA methylation may be necessary for normal growth from embryonic stages in mammals. When mutant embryonic stem cells lacking the murine DNA methyltransferase gene were introduced to a germline of mice they caused a recessive lethal phenotype. Methylation may also be linked to cancer development as methylation of tumor suppressor genes promotes tumorgenesis and metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen METTL7A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene METTL7A
Gene Accession No. Q9H8H3
Gene Alias 2210414H16Rik; 3300001H21Rik; Aam-B; methyltransferase like 7A; methyltransferase like 7A1; methyltransferase like 7A-like; methyltransferase-like protein 7A; METTL7A; Mettl7a1; PRO0066; Protein AAM-B; RGD1308407; UbiE1; UNQ1902/PRO4348
Gene Symbols METTL7A, Mettl7a1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-244 of human METTL7A (NP_0547523).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25840, 315306, 70152
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.