Learn More
Invitrogen™ METTL7B Polyclonal Antibody

Description
A suggested positive control is rat spleen tissue lysate. PA5-20637 can be used with blocking peptide PEP-0757.
Specifications
Specifications
| Antigen | METTL7B |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | METTL7B |
| Gene Accession No. | Q562C4, Q6UX53, Q9DD20 |
| Gene Alias | 0610006F02Rik; AI266817; ALDI; associated with lipid droplet protein 1; associated with lipid droplets 1; methyltransferase like 7B; methyltransferase-like protein 7B; Mettl7b; RGD1305205; UNQ594/PRO1180 |
| Gene Symbols | METTL7B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.