Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METTL7B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121075
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121075 Supplier Invitrogen™ Supplier No. PA5121075
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HT-29, Mouse liver, Rat liver. Immunogen sequence: RPGGVLFFWE HVAEPYGSWA FMWQQVFEPT WKHIGDGCCL TRETWKDLEN AQFSEIQMER QPPPLKWLPV GPHIMGKAVK.

MettL7B belongs to the methyltransferase superfamily. It is a probable methyltransferase. Methyltransferase is a type of transferase enzyme which transfers a methyl group from a donor to an acceptor. Often methylation occurs on nucleic bases in DNA or amino acids in protein structures. DNA methylation is often utilized to silence and regulate genes without changing the original DNA sequence. DNA methylation may be necessary for normal growth from embryonic stages in mammals. When mutant embryonic stem cells lacking the murine DNA methyltransferase gene were introduced to a germline of mice they caused a recessive lethal phenotype. Methylation may also be linked to cancer development as methylation of tumor suppressor genes promotes tumorgenesis and metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen METTL7B
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.544 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene METTL7B
Gene Accession No. Q562C4, Q6UX53, Q9DD20
Gene Alias 0610006F02Rik; AI266817; ALDI; associated with lipid droplet protein 1; associated with lipid droplets 1; methyltransferase like 7B; methyltransferase-like protein 7B; Mettl7b; RGD1305205; UNQ594/PRO1180
Gene Symbols METTL7B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 165-244 of human METTL7B (NP_689850.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196410, 366792, 71664
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.