Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METTL8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120734
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120734 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120734 Supplier Invitrogen™ Supplier No. PA5120734
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse liver, Rat testis. Immunogen sequence: MNMIWRNSIS CLRLGKVPHR YQSGYHPVAP LGSRILTDPA KVFEHNMWDH MQWSKEEEAA ARKKVKENSA VRVLLEEQVK YEREASKYWD TFYKIHKNKF FKDRNWLLRE FPEILPVDQK PEEKARESSW DHVKTSATNR.

TRUSTED_SUSTAINABILITY

Specifications

Antigen METTL8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.795 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene METTL8
Gene Accession No. A2AUU0, Q9H825
Gene Alias methyltransferase like 8; methyltransferase-like protein 8; METTL8; mRNA N(3)-methylcytidine methyltransferase METTL8; tension-induced/inhibited protein; TIP
Gene Symbols METTL8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human METTL8 (NP_001308089.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228019, 502633, 79828
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.