Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ METTL8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557265
Description
Immunogen sequence: MNMIWRNSIS CLRLGKVPHR YQSGYHPVAP LGSRILTDPA KVFEHNMWDH MQWSKEEEAA ARKKVKENSA VRVLLEE Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 82%.
Specifications
| METTL8 | |
| Polyclonal | |
| Unconjugated | |
| METTL8 | |
| methyltransferase like 8; methyltransferase-like protein 8; METTL8; mRNA N(3)-methylcytidine methyltransferase METTL8; tension-induced/inhibited protein; TIP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79828 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.34 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H825 | |
| METTL8 | |
| Recombinant protein corresponding to Human METTL8. Recombinant protein control fragment (Product #RP-96059). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction