Learn More
Mettler Toledo™ ErgoClips™ Solution Kits
Catalog No. 09320361
Click to view available options
For Use With (Equipment):
Analytical Balance
XP26 and XP56 Micro Balance
Includes:
Min weigh micro door, tube, micro flask, including
Minweigh door, flask, tube, small flask, round bot
Allow easy weighing directly into tare containers. Mettler Toledo ErgoClips Solution Kit includes all necessary tools for direct dosing with storage case.
  • Guarantees secure placement of different tare containers
  • Dosing is fast and safe thanks to the adjustable MinWeight Door
  • Accurate weighing for reliable, complete sample transfer
Solution Kit Contents:
  • ErgoClip Flask
  • ErgoClip Tube
  • ErgoClip Flask (Small)
  • ErgoClip Round Bottom Flask
  • ErgoClip Basket (Small)
  • ErgoClip Weighing Boat
  • ErgoClip Titration Basket
  • ErgoClip Quantos and vial adapters
  • ErgoClip Filter holder
  • MinWeight Door
  • Stand
  • Various tare containers
  • Spatula
  • Tweezers
Solution Kit Micro Contents:
  • MinWeight Door micro
  • ErgoClip Tube (without holder)
  • ErgoClip Flask micro, including extension
  • ErgoClip Basket micro
  • Stand for ErgoClips micro
  • SmartGrid
  • Tweezers
  • 5mL volumetric flasks
  • Reagent tube, small
  • 1.5 and 0.5mL centrifuge tube
  • 2mL HPLC tube
  • Micro spatula
Solution kit for use with analytical balances.
Micro kit for use with micro balances XP26 or XP56.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Analytical Balance
Includes Minweigh door, flask, tube, small flask, round bot
Type ErgoClip™ Solution Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.