Learn More
Mettler Toledo™ InLab™ Conductivity Sensors Available on GSA/VA Contract for Federal Government customers only.
Sensors with Intelligent Sensor Management (ISM) technology. Mettler Toledo InLab Conductivity Sensors with integrated NTC 30kOhm temperature sensor enables automatic temperature compensation.
Manufacturer:  Mettler Toledo™ 51344114
 View more versions of this product
Catalog No. 01-917-150
Add to cart
Description
Description
  • High-performance conductivity cells with fixed cable (LTW)
  • Waterproof to IP67, ideal for field measurements
  • Easy to maintain
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
0 to 100°C
Mini DIN (IP67)
12mm
4.72 in.
120mm
Epoxy shaft/4 Graphite poles
Dry
Conductivity
InLab 738-ISM-10m
0.57cm-1
0.47 in.
SevenGo Duo™ SG23 and SevenGo Duo pro™ SG78 Dual Channel Portable Meters
393.7 in.
10m
0.01 to 1000μS/cm
0° to 100°C
NTC 30kilohm