Learn More
Mettler Toledo™ MP Portable pH Meters
Shop All METTLER TOLEDO ProductsDescription
Includes
All packages include pH meter, batteries, instructions and an instrument sealing kit. The B (basic) package offers the meter without an electrode. The BLE package includes a low-cost, 3-in-1 non-IP67 electrode (01-911-250) and pH buffer sachets; the BE package includes an InLab™ 413 combination pH electrode with integrated temperature sensor and pH buffer sachets. Field Kits (FK) include the InLab combination electrode, pH buffer sachets, and a carrying case.
pH and pH GLP models; Fully waterproofed to IP67 specifications.
Rugged and durable for use in both lab and field environments—so reliable, a battery change every 500 hours is the only internal maintenance they'll ever need. Ultrasonically welded casings are fully waterproofed to IP67 specifications—when used with special booted electrodes, the units are dustproof and watertight to a depth of 1m for 30 minutes.
MP120. Basic pH meter is an excellent value—features pH range of 0.00 to 14.00, electrode condition and slope % indicators, selectable compensated buffers, auto-endpoint and auto-off functions and stable-reading indicator.
MP125. Expands on the capabilities of the MP120 with a pH range of -2.00 to 16.00 and enhanced GLP features including datalogging, date and time stamping, buffer prompting, 0- to 4-hour calibration reminder and last calibration information (including E0 and slope values). Clear visual icons display various functions including stable reading, auto endpoint, date/time, selectable buffers, serial data output, auto-off override and probe condition (immediately after calibration).
Specifications
Specifications
| Includes | 3-in-1 Electrode |
| Type | Portable pH Meter |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.