Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mettler Toledo™ Pump Button for MettlerToledo Karl Fischer™ Titrators
Description
- For solvent filling/evacuation
- For model DL31/38 Karl Fischer titrators

Specifications
Specifications
Color | Blue |
Type | Pump Button |
For Use With (Application) | Solvent filling/evacuation |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction