Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEX3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572749
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572749 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572749 Supplier Invitrogen™ Supplier No. PA572749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MEX3B Polyclonal Antibody for Western Blot, ICC/IF, ELISA

RNA-binding protein. May be involved in post-transcriptional regulatory mechanisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEX3B
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MEX3B
Gene Accession No. Q69Z36, Q6ZN04
Gene Alias 4931439A04Rik; KIAA2009; M-04814; mex3 homolog B; mex3 homolog B (C. elegans); mex-3 RNA binding family member B; MEX3B; MEX-3B; mKIAA2009; ring finger and KH domain containing 3; RING finger and KH domain-containing protein 3; RING finger protein 195; RKHD3; RNA-binding protein MEX3B; RNF195
Gene Symbols MEX3B
Host Species Rabbit
Immunogen RKHD3 antibody for a 16 amino acid synthetic peptide from near the center of human RKHD3. The immunogen is within amino acids 180 - 230 of RKHD3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108797, 308790, 84206
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.