Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-N-butylammonium tetrafluoroborate, Thermo Scientific™

Catalog No. AC358700250
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12107-76-5
Molecular Formula C8H20BF4N
Molecular Weight (g/mol) 217.06
MDL Number MFCD00034899
InChI Key CAZKFGXFAVXKIF-UHFFFAOYSA-O
Synonym di-n-butylammonium tetrafluoroborate, dibutylazanium tetrafluoroborate, dibutylamine tetrafluoroborate, dibutylammonium tetrafluoroborate 1-, dibutylammonium tetrafluoroborate, dibutylammonium, tetrafluoroborate, dibutylamine, tetrafluoroborate 1-, 1-butanamine, n-butyl-, tetrafluoroborate 1-, dibutylamine tetrafluoborate, c8h20n.bf4
PubChem CID 25510
IUPAC Name dibutylazanium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CCCC[NH2+]CCCC

Specifications

Melting Point 266.0°C
Color White
Elemental Analysis 96% min. ( ex B ), 96% min. ( ex F )
Infrared Spectrum Authentic
Packaging Glass Bottle
Linear Formula (CH3CHCH2CH3)2NH2BF4
Quantity 25 g
Formula Weight 216.95
Physical Form Powder or Flakes
Chemical Name or Material Di-N-butylammonium tetrafluoroborate
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.