Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MFAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139758
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139758 Supplier Invitrogen™ Supplier No. PA5139758
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: KELEENKRSL AALDALNTDD ENDEEEYEAW KVRELKRIKR DREDREALEK EKAEIERMRN LTEEERRAEL RANGKVITNK AVKGKYKFLQ KYYHRGAFFM DEDEEVYKRD FSAPTLEDHF NKTILPKVMQ VKNFGRSG Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 100%.

MFAP1 is a component of the elastin-associated microfibrils. Microfibrils, found either in association with elastin or independently, are an important component of the extracellular matrix of many tissues. MFAP1 is one of a number of proteins demonstrated to be essential for cell division.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFAP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MFAP1
Gene Accession No. P55081
Gene Alias 4432409M24Rik; Mfap1; Mfap1a; Mfap1b; microfibril associated protein 1; microfibrillar associated protein 1; microfibrillar-associated protein 1; microfibrillar-associated protein 1A; Microfibrillar-associated protein 1B; microfibrillar-associated protein 1-like protein; RGD1562232; Spliceosome B complex protein MFAP1; Spliceosome B complex protein MFAP1A; Spliceosome B complex protein MFAP1B
Gene Symbols MFAP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MFAP1. Recombinant protein control fragment (Product #RP-90794).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4236
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.