Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFAP1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS7810R
Description
MFAP1 is a component of the elastin-associated microfibrils. Microfibrils, found either in association with elastin or independently, are an important component of the extracellular matrix of many tissues. MFAP1 is one of a number of proteins demonstrated to be essential for cell division.Specifications
| MFAP1 | |
| Polyclonal | |
| Unconjugated | |
| MFAP1 | |
| 4432409M24Rik; BOS_20487; MFAP1; Mfap1a; Mfap1b; microfibril associated protein 1; microfibrillar associated protein 1; Microfibrillar-associated protein 1; Microfibrillar-associated protein 1A; Microfibrillar-associated protein 1B; RGD1562232; Spliceosome B complex protein MFAP1; Spliceosome B complex protein MFAP1A; Spliceosome B complex protein MFAP1B | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4236 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
| P55081 | |
| MFAP1 | |
| KLH conjugated synthetic peptide derived from human MFAP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction