Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MFAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547680
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547680 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547680 Supplier Invitrogen™ Supplier No. PA547680
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 35% cross-reactivity with recombinant human MAGP-1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Microfibrillar-associated protein 2 is a major antigen of elastin-associated microfibrils and a candidate for involvement in the etiology of inherited connective tissue diseases. Four transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFAP2
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene MFAP2
Gene Accession No. P55002
Gene Alias AI893631; Magp; Magp1; MAGP-1; MAGP-1FLJ50901; MAGPMFAP-2; MFAP2; MFAP-2; microfibril associated protein 2; microfibril-associated glycoprotein; microfibril-associated glycoprotein 1; microfibrillar associated protein 2; microfibrillar-associated protein 2; Tropoelastin-binding protein
Gene Symbols MFAP2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MAGP-1/MFAP2 Gln17-Cys183.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17150
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.