Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MFAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552425
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552425 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552425 Supplier Invitrogen™ Supplier No. PA552425
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QYTHYSDQID NPDYYDYQEV TPRPSEEQFQ FQSQQQVQQE VIPAPTPEPG NAELEPTEPG PLDCREEQYP CTRLYSIHRP CKQCLNEVCF YSLRRVYVIN KEICVRTVCA HEELLRADLC RDKFSKCG Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.

Microfibrillar-associated protein 2 is a major antigen of elastin-associated microfibrils and a candidate for involvement in the etiology of inherited connective tissue diseases. Four transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFAP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MFAP2
Gene Accession No. P55001
Gene Alias AI893631; Magp; Magp1; MAGP-1; MAGP-1FLJ50901; MAGPMFAP-2; MFAP2; MFAP-2; microfibril associated protein 2; microfibril-associated glycoprotein; microfibril-associated glycoprotein 1; microfibrillar associated protein 2; microfibrillar-associated protein 2; Tropoelastin-binding protein
Gene Symbols MFAP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MFAP2. Recombinant protein control fragment (Product #RP-102185).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4237
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.