Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MFAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587728
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587728 Supplier Invitrogen™ Supplier No. PA587728
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGQYDLDPLP PFPDHVQYTH YSDQIDNPDY YDYQEVTPRP SEEQFQFQSQ QQVQQEVIPA PTPEPGNAEL EPTEPGPLDC REEQYPCTRL YSIHRPCKQC LNEVCFYSLR RVYVINKEIC VRTVCAHEEL LRADLCRDKF SKCGVMASSG LCQSVAASCA RSCGSC; Positive Samples: HeLa, OVCAR3, MCF7, Mouse lung, Mouse brain, Rat lung; Cellular Location: extracellular matrix, extracellular space.

Microfibrillar-associated protein 2 is a major antigen of elastin-associated microfibrils and a candidate for involvement in the etiology of inherited connective tissue diseases. Four transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFAP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MFAP2
Gene Accession No. P55001, P55002
Gene Alias AI893631; Magp; Magp1; MAGP-1; MAGP-1FLJ50901; MAGPMFAP-2; MFAP2; MFAP-2; microfibril associated protein 2; microfibril-associated glycoprotein; microfibril-associated glycoprotein 1; microfibrillar associated protein 2; microfibrillar-associated protein 2; Tropoelastin-binding protein
Gene Symbols MFAP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 18-183 of human MFAP2 (NP_0594531).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17150, 313662, 4237
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.