Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MFAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544660
Description
Peptide sequence: MRAAYLFLLF LPAGLLAQGQ YDLDPLPPFP DHVQYTHYSD QIDNPDYYDY Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
Microfibrillar-associated protein 2 is a major antigen of elastin-associated microfibrils and a candidate for involvement in the etiology of inherited connective tissue diseases. Four transcript variants encoding two different isoforms have been found for this gene.
Specifications
MFAP2 | |
Polyclonal | |
Unconjugated | |
MFAP2 | |
AI893631; Magp; Magp1; MAGP-1; MAGP-1FLJ50901; MAGPMFAP-2; MFAP2; MFAP-2; microfibril associated protein 2; microfibril-associated glycoprotein; microfibril-associated glycoprotein 1; microfibrillar associated protein 2; microfibrillar-associated protein 2; Tropoelastin-binding protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
4237 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P55001 | |
MFAP2 | |
Synthetic peptide directed towards the N-terminal of human MFAP2 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction