Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFAP3 Antibody (OTI1B3), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272657JF646
Description
MFAP3 Monoclonal specifically detects MFAP3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
MFAP3 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MFAP3 | |
Full length recombinant protein of human MFAP3 (NP_005918) produced in HEK293T cells. | |
0.1 mL | |
4238.0 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1B3 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
DKFZp586F0219, microfibril-associated glycoprotein 3, microfibrillar-associated protein 3 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction