Learn More
Invitrogen™ MFAP3 Monoclonal Antibody (1C7)

Description
Peptide Sequence: DDPDDLGERI KERPALNAQG GIYVINPEMG RSNSPGGDSD DGSLNEQGQE IAVQVSVHLQ SETKSIDTES QGSSHFSPPD DIGSAESNCN YKDGAYENCQ L.
Specifications
Specifications
| Antigen | MFAP3 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1C7 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | MFAP3 |
| Gene Accession No. | P55082 |
| Gene Alias | MFAP3; Microfibril-associated glycoprotein 3; microfibrillar associated protein 3; microfibrillar-associated protein 3 |
| Gene Symbols | MFAP3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.