Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFAP3 Monoclonal Antibody (OTI1G1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA506943

Description
Component of the elastin-associated microfibrils.Specifications
MFAP3 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P55082 | |
MFAP3 | |
Full length human recombit protein of human MFAP3 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1G1 | |
Unconjugated | |
MFAP3 | |
microfibril associated protein 3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4238 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction