Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MFAP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565548
Description
Immunogen sequence: DVDFDQMVSLE ANRSSYNASF PSSFELSASS HSDDDVIIAK EGTSVSIECL LTASHYEDVH WHNSKGQQLD GRSRGGKWLV SDNFLNITNV AFDDRGLYTC FVTSPIRASY SVTLRVI Highest antigen sequence identity to the following orthologs - mouse 73%, rat 74%.
Component of the elastin-associated microfibrils.
Specifications
MFAP3 | |
Polyclonal | |
Unconjugated | |
MFAP3 | |
MFAP3; Microfibril-associated glycoprotein 3; microfibrillar associated protein 3; microfibrillar-associated protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4238 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P55082 | |
MFAP3 | |
Recombinant Human MFAP3. Recombinant protein control fragment (Product #RP-106220). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction