Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFAP3L Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986329
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6329 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6329 Supplier Bioss Supplier No. BS5101R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MFAP3L is a protein coding gene. May participate in the nuclear signaling of EGFR and MAPK1/ERK2. May a have a role in metastasis.

Specifications

Antigen MFAP3L
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MFAP3L
Gene Accession No. O75121
Gene Alias 4933428A15Rik; 5430405D20Rik; AI461995; AW125052; BOS_8565; HSD39; HSD-39; KIAA0626; Mfap3l; microfi brillar-associated protein 3-like; microfibril associated protein 3 like; microfibrillar associated protein 3 like; Microfibrillar-associated protein 3-like; mKIAA0626; NYD-sp9; Testis development protein NYD-SP9
Gene Symbols MFAP3L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MFAP3L.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9848
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.