Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFAP4 Antibody (12D11), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242638AF647
Description
MFAP4 Monoclonal specifically detects MFAP4 in Human samples. It is validated for Western Blot, ELISA.Specifications
MFAP4 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MFAP4 | |
Recombinant human MFAP4 (22-255aa) purified from E. coli | |
100 μL | |
Signal Transduction | |
4239 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
12D11 | |
Western Blot, ELISA | |
microfibril-associated glycoprotein 4, microfibrillar-associated protein 4 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction