Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFAP4 Antibody (12D11), DyLight 650, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242638C

Catalog No. NBP242638C


Only null left
Add to Cart

Description

Description

MFAP4 Monoclonal specifically detects MFAP4 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

MFAP4
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
MFAP4
Recombinant human MFAP4 (22-255aa) purified from E. coli
100 μL
Signal Transduction
4239
Store at 4C in the dark.
IgG1 κ
Western Blot, ELISA
12D11
Western Blot, ELISA
microfibril-associated glycoprotein 4, microfibrillar-associated protein 4
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.