Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MFAP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100280
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100280 Supplier Invitrogen™ Supplier No. PA5100280
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MFAP4.

MFAP4 is a protein with similarity to a bovine microfibril-associated protein. The protein has binding specificities for both collagen and carbohydrate. It is thought to be an extracellular matrix protein which is involved in cell adhesion or intercellular interactions. The gene is located within the Smith-Magenis syndrome region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFAP4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MFAP4
Gene Accession No. P55083, Q9D1H9
Gene Alias 1110007F23Rik; 36 kDa MAP; 36 kDa microfibril-associated glycoprotein; Magp-36; MFAP4; microfibril associated protein 4; microfibril-associated glycoprotein 4; microfibrillar associated protein 4; microfibrillar-associated protein 4; Unknown (protein for MGC:137920)
Gene Symbols MFAP4
Host Species Rabbit
Immunogen A synthesized peptide derived from human MFAP4(Accession P55083), corresponding to amino acid residues C18-V68.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287382, 4239, 76293
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.