Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MFAP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552706
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552706 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552706 Supplier Invitrogen™ Supplier No. PA552706
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVNSQRGDDV TQATPETFTE DPNLVNDPAT DETVLAVLAD IAPSTDDLAS LSEKNTTAEC WDEKFTCTRL YSVHRPVKQC IHQLCFTSLR RMYIVNKEIC SRLVCKEHEA MKDELCRQMA GLPPRRLRRS NYFRLPPCEN V Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 83%.

This gene encodes a 25-kDa microfibril-associated glycoprotein which is rich in serine and threonine residues. It lacks a hydrophobic carboxyl terminus and proline-, glutamine-, and tyrosine-rich regions, which are characteristics of a related 31- kDa microfibril-associated glycoprotein (MFAP2). The close similarity between these two proteins is confined to a central region of 60 aa where precise alignment of 7 cysteine residues occurs. The structural differences suggest that this encoded protein has some functions that are distinct from those of MFAP2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFAP5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MFAP5
Gene Accession No. Q13361
Gene Alias AAT9; MAGP2; MAGP-2; MFAP5; MFAP-5; Microfibril-associated glycoprotein 2; microfibril-associated glycoprotein-2; microfibrillar associated protein 5; microfibrillar-associated protein 5; MP25; THE1A-MFAP5
Gene Symbols MFAP5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MFAP5. Recombinant protein control fragment (Product #RP-89743).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8076
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.