Learn More
Promega mFcgammaRIV ADCC Bioassay Effector Cells, Propagation Model, 1 each
Catalog No. PRM1212 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Each
Catalog No. Quantity
PRM1212 1 Each
1 options
Catalog No. PRM1212 Supplier Promega Supplier No. M1212
Add to Cart
Add to Cart
Request a Quote
The mFcγRIV ADCC Bioassay Effector Cells, Propagation Model, provides a biologically relevant and specific MOA-based measure of mFcγRIV ADCC without the complex workflow and variability inherent in primary cell-based assays.
  • Allows Propagation and Banking of mFcγRIV ADCC Bioassay Effector Cells
Specifications
Content And Storage BELOW -140°C
Quantity 1 Each