Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFF Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318442AF532
Description
MFF Polyclonal antibody specifically detects MFF in Human samples. It is validated for Western BlotSpecifications
MFF | |
Polyclonal | |
Western Blot | |
mitochondrial fission factor | |
Recognizes region between residues Between Between 50 and 100.. | |
0.1 mL | |
Apoptosis, Cell Biology, Endocrinology | |
56947 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction