Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mff, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABS208725UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS208725UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS208725UL Supplier MilliporeSigma Supplier No. ABS208725UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Mff, Cat. No. ABS2087, is a rabbit polyclonal antibody that detects Mitochondrial fission factor (Mff) and has been tested for use in Immunocytochemistry, Immunofluorescence, and Western Blotting.

Mitochondrial fission factor (UniProt: Q9GZY8; also known as Mff) is encoded by the MFF (also known as C2orf33, AD030, AD033, GL004) gene (Gene ID: 56947) in human. Mff is a single-pass type IV membrane protein that is highly expressed in heart, kidney, liver, brain, muscle, and stomach. Mff is a homodimeric protein, but may also interact with other proteins. It plays a role in mitochondrial and peroxisomal fission and promotes the recruitment and association of the fission mediator dynamin-related protein 1 (DNM1L) to the mitochondrial surface. It is also suggested to be involved in regulation of synaptic vesicle membrane dynamics by recruitment of DNM1L to clathrin-containing vesicles. Mff has a cytoplasmic domain (aa 1-322), a helical domain (aa 323-340), and a mitochondrial intermembrane domain (aa 341-342). Mff is shown to be an essential factor for mitochondrial recruitment of Drp1 during mitochondrial fission in mammalian cells and its down-regulation inhibits mitochondrial recruitment of Drp1 and induces mitochondria elongation. Also, exogenous expression of Mff is shown to induce extensive mitochondrial fission. Mutations in MFF gene can cause a form of encephalopathy that is characterized by delayed psychomotor development, severe hypotonia with inability to walk, and microcephaly. (Ref.: Otera, H., et al. (2010). J. Cell Biol. 191 (6); 1141 1158; Gandre-Babbe, S., and van der Bliek, AM. (2008). Mol. Biol. Cell. 19(6): 2402-2412).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mff
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representative lot detected Mff in Western Blotting applications (Mishra, P., et. al. (2015). Cell Metab. 22(6):1033-44).Immunofluorescence Analysis: A representative lot detected Mff in Immunofluorescence applications (Mishra, P., et. al. (2015). Cell Metab. 22(6):1033-44).
Formulation Rabbit polyclonal antiserum with 0.05% sodium azide.
Gene Alias Mitochondrial fission factor
Gene Symbols MFF;C2orf33;AD030;AD033;GL004
Host Species Rabbit
Immunogen His-taged full length human recombinant mitochondrial fission factor (Mff).
Molecular Weight of Antigen 38.47 kDa calculated.
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects mitochondrial fission factor (Mff) in human cells.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.