Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFG-E8 Antibody (SPM291), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00 - $524.00

Specifications

Antigen MFG-E8
Clone SPM291
Concentration 0.2mg/mL
Dilution Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP24441100
SDP
View Documents
Novus Biologicals
NBP2444110.02MG
0.02 mg
Each for $268.00
Only null left
Add to Cart
 
NBP24441101
SDP
View Documents
Novus Biologicals
NBP2444110.1MG
0.1 mg
Each for $524.00
Only null left
Add to Cart
 
NBP24441102
SDP
View Documents
Novus Biologicals
NBP2444110.2MG
0.2 mg N/A N/A N/A
Description

Description

Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

MFG-E8 Monoclonal specifically detects MFG-E8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

MFG-E8
0.2mg/mL
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
Mouse
Cancer
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
4240
Human milk fat globule membrane preparation
Primary
Store at 4C.
45 kDa
SPM291
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml
Monoclonal
Purified
RUO
Human
BA46, Breast epithelial antigen BA46, EDIL1, hP47, HsT19888, lactadherin, medin, MFG1, MFG-E8, MFGM, Milk fat globule-EGF factor 8, milk fat globule-EGF factor 8 protein, O-acetyl disialoganglioside synthase, OAcGD3S, SED1, SPAG10, sperm associated antigen 10, sperm surface protein hP47
MFGE8
IgG1 κ
Protein A or G purified
Recognizes a protein of 40-45kDa, identified as human milk fat globule membrane protein (HMFG). HMFG is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as to the outer surface of the human milk fat globule. HMFG is considered as a differentiation marker. It is useful as specific breast epithelial marker and can also provide a tool to study the role of the cell surface in normal and neoplastic mammary development.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.