Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFGE8 (Lactadherin) Monoclonal Antibody (EDM45), Invitrogen™

Catalog No. PIMA513556
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513556 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513556 Supplier Thermo Scientific Supplier No. MA513556
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-13556 targets Human Milk Fat Globule 1 in IF, IHC (P), IP, and WB applications and shows reactivity with Human samples. The MA5-13556 immunogen is delipidated human milk fat globule membrane preparation.

This gene encodes a preproprotein that is proteolytically processed to form multiple protein products. The major encoded protein product, lactadherin, is a membrane glycoprotein that promotes phagocytosis of apoptotic cells. This protein has also been implicated in wound healing, autoimmune disease, and cancer. Lactadherin can be further processed to form a smaller cleavage product, medin, which comprises the major protein component of aortic medial amyloid (AMA). Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFGE8 (Lactadherin)
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone EDM45
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene MFGE8
Gene Accession No. Q08431
Gene Alias AA408458; AGS; AI325141; BA46; breast epithelial antigen BA46; EDIL1; EGF/factor VIII; HMFG; hP47; HsT19888; lactadherin; Lactadherin short form; Medin; Mfge8; MFG-E8; MFGM; MFGME8; MFGMP-E8; milk fat globule glycoprotein; milk fat globule membrane; milk fat globule membrane protein; Milk fat globule-EGF factor 8; milk fat globule-EGF factor 8 protein; MP47; O-acetyl disialoganglioside synthase; O-acetyl GD3 ganglioside synthase; O-acetyltransferase milk fat globule membrane protein; O-acetyltransferase Milk fat glolbule membrane protein; OAcGD3S; P47; SED1; SPAG10; sperm associated antigen 10; sperm surface protein hP47; sperm surface protein SP47
Gene Symbols MFGE8
Host Species Mouse
Immunogen Delipidated human milk fat globule membrane preparation
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4240
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.