Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFGE8 Mouse anti-Human, Clone: OTI7A4, liquid, TrueMAB™
SDP

Catalog No. 501682684
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2684 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2684 Supplier OriGene Supplier No. TA809279
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lactadherin reacts with 40-45 kDa glycoprotein in human milk fat globule membrane preparation.

Specifications

Antigen MFGE8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MFGE8
Gene Accession No. Q08431
Gene Alias BA46, EDIL1, HMFG, HsT19888, MFG-E8, MFGM, OAcGD3S, SED1, SPAG10, hP47
Gene Symbols MFGE8
Host Species Mouse
Immunogen Full length human recombit protein of human MFGE8 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4240
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.