Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MFHAS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565877
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565877 Supplier Invitrogen™ Supplier No. PA565877
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RVGLWKIKDNP LIQPPYEVCM KGIPYIAAYQ KELAHSQPAV QPRLKLLLMG HKAAGKTLLR HCLTEERVEG CPGGGDKEKC YPPSP Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.

Identified in a human 8p amplicon, this gene is a potential oncogene whose expression is enhanced in some malignant fibrous histiocytomas (MFH). The primary structure of its product includes an ATP/GTP-binding site, three leucine zipper domains, and a leucine-rich tandem repeat, which are structural or functional elements for interactions among proteins related to the cell cycle, and which suggest that overexpression might be oncogenic with respect to MFH.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFHAS1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MFHAS1
Gene Accession No. Q9Y4C4
Gene Alias leucine rich repeat containing 65; LRRC65; malignant fibrous histiocytoma amplified sequence 1; malignant fibrous histiocytoma-amplified sequence 1; Malignant fibrous histiocytoma-amplified sequence with leucine-rich tandem repeats 1; malignant fibrous histiocytoma-amplified sequences with leucine-rich tandem repeats 1; MASL1; MFH-amplified sequences with leucine-rich tandem repeats 1; MFHAS1
Gene Symbols MFHAS1
Host Species Rabbit
Immunogen Recombinant Human MFHAS1. Recombinant protein control fragment (Product #RP-106547).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9258
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.