Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MFI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532920
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532920 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532920 Supplier Invitrogen™ Supplier No. PA532920
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset, Mouse, Dog, Elephant, Panda, Horse, Rabbit (100%) Bovine, Hamster, Pig (93%).

The protein encoded by this gene is a cell-surface glycoprotein found on melanoma cells. The protein shares sequence similarity and iron-binding properties with members of the transferrin superfamily. The importance of the iron binding function has not yet been identified. This gene resides in the same region of chromosome 3 as members of the transferrin superfamily. Alternative splicing results in two transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFI2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene MELTF
Gene Accession No. O97490, P08582, Q9R0R1
Gene Alias antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and 96.5; CD228; MAP97; Melanoma-associated antigen p97; melanoma-associated antigen p97, isoform 2; melanotransferrin; MELTF; membrane-bound transferrin-like protein; membrane-bound transferrin-like protein p97; Mfi2; Mtf; MTF1; p97
Gene Symbols MELTF
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from N-terminus of human MFI2.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100008603, 100060123, 30060, 4241, 488035
Target Species Human, Mouse, Canine, Rabbit, Monkey, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.