Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MFI2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552127
Description
Immunogen sequence: RSEDYELLCP NGARAEVSQF AACNLAQIPP HAVMVRPDTN IFTVYGLLDK AQDLFGDDHN KNGFKMFDSS NYHGQDLLFK DATVRAVPVG EKTTYRGWLG LDYVAALEGM SSQQC Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.
The protein encoded by this gene is a cell-surface glycoprotein found on melanoma cells. The protein shares sequence similarity and iron-binding properties with members of the transferrin superfamily. The importance of the iron binding function has not yet been identified. This gene resides in the same region of chromosome 3 as members of the transferrin superfamily. Alternative splicing results in two transcript variants.
Specifications
MFI2 | |
Polyclonal | |
Unconjugated | |
MELTF | |
antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and 96.5; CD228; MAP97; Melanoma-associated antigen p97; melanoma-associated antigen p97, isoform 2; melanotransferrin; MELTF; membrane-bound transferrin-like protein; membrane-bound transferrin-like protein p97; Mfi2; Mtf; MTF1; p97 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4241 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P08582 | |
MELTF | |
Recombinant protein corresponding to Human MFI2. Recombinant protein control fragment (Product #RP-102453). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction