Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFN1, Mouse anti-Human, Clone: 3C9, Abnova™

Catalog No. 89335035 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-035 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-035 Supplier Abnova Corporation Supplier No. H00055669M04J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sequence: MAEPVSPLKHFVLAKKAITAIFDQLLEFVTEGSHFVEATYKNPELDRIATEDDLVEMQGYKDKLSIIGEVLSRRHMKVAFFGRTSSGKSSVINAMLWDKVLPSGIGHITNCFLSVEGTDGDKAYLMTEGSDEKKSVKTVNQLAHALHMDKDLKAGCLVRVFWPKAKCALLRDDLVLVDSPGTDVTTELDSWIDKFCLDADVFVLVANSESTLMNTEKHFFHKVNERLSKPNIFILNNRWDASASEPEYMEDVRRQHMERCLHFLVEELKVVNALEAQNRIFFVSAKEVLSARKQKAQGMPESGVALAEGFHARLQEFQNFEQIFEECISQSAVKTKFEQHTIRAKQILATVKNIMDSVNLAAEDKRHYSVEEREDQIDRLDFIRNQMNLLTLDVKKKIKEVTEEVANKVSCAMTDEICRLSVLVDEFCSEFHPNPDVLKIYKSELNKHIEDGMGRNLADRCTDEVNALVLQTQQEIIENLKPLLPAGIQDKLHTLIPCKKFDLSYNLNYHKLCSDFQEDIVFRFSLGWSSLVHRFLGPRNAQRVLLGLSEPIFQLPRSLASTPTAPTTPATPDNASQEELMITLVTGLASVTSRTSMGIIIVGGVIWKTIGWKLLSVSLTMYGALYLYERLSWTTHAKERAFKQQFVNYATEKLRMIVSSTSANCSHQVKQQIATTFARLCQQVDITQKQLEEEIARLPKEIDQLEKIQNNSKLLRNKAVQLENELENFTKQFLPSSNEES

Specifications

Antigen MFN1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3C9
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene mitofusin 1
Gene Accession No. BC040557
Gene Alias DKFZp762F247/FLJ20693/MGC41806/hfzo1/hfzo2
Gene Symbols MFN1
Host Species Mouse
Immunogen MFN1 (AAH40557, 1 a.a. ∼ 741 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55669
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.