Learn More
Invitrogen™ MFN1 Polyclonal Antibody

Description
Positive test controls include: PC-3, Mouse kidney, Rat kidney. The target is usually found in the following locations: Cytoplasm, Mitochondrion outer membrane, Multi-pass membrane protein. Immunogen sequence: MAETVSPLKH FVLAKKAITA IFGQLLEFVT EGSHFVEATY RNPELDRIAS EDDLVEIQGY RNKLAVIGEV LSRRHMKVAF FGRTSSGKSS VINAMLWDKV.
Specifications
Specifications
| Antigen | MFN1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.42 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MFN1 |
| Gene Accession No. | Q811U4, Q8IWA4, Q8R4Z9 |
| Gene Alias | 2310002F04Rik; 6330416C07Rik; D3Ertd265e; DKFZp762F247; EC 3.6.5; FLJ20693; Fzo homolog; Fzo1b; hfzo1; hfzo2; HR2; Mfn1; MFN2; MGC41806; mitochondrial mitofusin 1; mitochondrial transmembrane GTPase Fzo-1; mitochondrial transmembrane GTPase FZO1B; mitochondrial transmembrane GTPase FZO-2; mitofusin 1; mitofusin 2; mitofusin-1; mKIAA4032; putative transmembrane GTPase; transmembrane GTPase MFN1 |
| Gene Symbols | MFN1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.