Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MFNG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142901
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142901 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142901 Supplier Invitrogen™ Supplier No. PA5142901
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize isoform 1 (NP_002396.2). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.

This gene is a member of the fringe gene family which also includes radical and lunatic fringe genes. They all encode evolutionarily conserved secreted proteins that act in the Notch receptor pathway to demarcate boundaries during embryonic development. While their genomic structure is distinct from other glycosyltransferases, fringe proteins have a fucose-specific beta-1,3-N-acetylglucosaminyltransferase activity that leads to elongation of O-linked fucose residues on Notch, which alters Notch signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFNG
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene MFNG
Gene Accession No. O00587
Gene Alias AW546563; beta-1,3-N-acetylglucosaminyltransferase manic fringe; EC 2.4.1.222; manic fringe homolog; Mfng; MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase; O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Symbols MFNG
Host Species Goat
Immunogen Peptide with sequence C-NRPIHASEPQPHNRT.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4242
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.