Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MFRP Monoclonal Antibody (291204)

Rat Monoclonal Antibody

Supplier:  Invitrogen™ MA523996

Catalog No. PIMA523996


Only null left
Add to Cart

Description

Description

In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) Frizzled-1, -2, -3, -4, -7, -8, -9, recombinant human Frizzled-5, -6, -10, or rmMFRP is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

MFRP is a member of the frizzled-related proteins. It may play a role in eye development, as mutations in this gene have been associated with nanophthalmos, posterior microphthalmia, retinitis pigmentosa, foveoschisis, and optic disc drusen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MFRP
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q9BY79
MFRP
Mouse myeloma cell line NS0-derived recombinant human MFRP Ser101-Pro579.
100 μg
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin)
291204
Unconjugated
MFRP
C1QTNF5; DKFZp586B0621; FLJ30570; MCOP5; Membrane frizzled-related protein; membrane-type frizzled-related protein; MFRP; MGC32938; NNO2; rd6
Rat
Protein A/G
RUO
83552
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.