Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MFRP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547490
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547490 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547490 Supplier Invitrogen™ Supplier No. PA547490
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human (rh) MFRP is observed, 15% cross-reactivity with recombinant mouse (rm) Frizzled-4 is observed, 10% cross-reactivity with rmFrizzled-1, rhFrizzled-5, rmFrizzled-8, and rmFrizzled-10 is observed, and 5% cross-reactivity with rmFrizzled-3, rmFrizzled-6, and rmFrizzled-7 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

MFRP is a member of the frizzled-related proteins. It may play a role in eye development, as mutations in this gene have been associated with nanophthalmos, posterior microphthalmia, retinitis pigmentosa, foveoschisis, and optic disc drusen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFRP
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene MFRP
Gene Accession No. Q8K480
Gene Alias C1QTNF5; DKFZp586B0621; FLJ30570; MCOP5; Membrane frizzled-related protein; membrane-type frizzled-related protein; MFRP; MGC32938; NNO2; rd6
Gene Symbols MFRP
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MFRP Gln91-Pro584.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259172
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.