Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFSD10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501727230 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7230 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7230 Supplier Proteintech Group Inc Supplier No. 115181AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TETRAN′s overexpression in cultured human cells caused resistance to some NSAIDs (Non-steroidal anti-inflammatory drugs), suggesting that TETRAN is an efflux pump for some NSAIDs.

Specifications

Antigen MFSD10
Applications Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MFSD10
Gene Accession No. Q14728, Q9D2V8
Gene Alias MFSD10, TETRAN, TETTRAN
Gene Symbols Mfsd10
Host Species Rabbit
Immunogen MFSD10 Fusion Protein Ag2080
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10227, 305449, 68294
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.