Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFSD11 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306336APC
Description
MFSD11 Polyclonal antibody specifically detects MFSD11 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
MFSD11 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
ET, FLJ20226, FLJ22196, major facilitator superfamily domain containing 11, major facilitator superfamily domain-containing protein 11, protein ET, UNC93-like protein MFSD11 | |
Produced in rabbits immunized with E. coli-derived Human MFSD11 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
79157 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction