Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFSD2B Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281737AF750
This item is not returnable.
View return policy
Description
MFSD2B Polyclonal antibody specifically detects MFSD2B in Human, Rat samples. It is validated for Western Blot, ELISASpecifications
MFSD2B | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot, ELISA | |
Alexa Fluor 750 | |
MFSD2B major facilitator superfamily domain containing 2B | |
MFSD2B antibody was raised against a 15 amino acid synthetic peptide near the center of human MFSD2B. The immunogen is located within amino acids 240 - 290 of MFSD2B. Amino Acid Squence: VKERPDPSAPASGPG | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
388931 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction