Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MFSD2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560628
Description
Immunogen sequence: VGSTPKTPSR DASSRLSLRR RTSYSL Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 85%.
Multidrug transporters, such as MFSD2B, are membrane proteins that expel a wide spectrum of cytotoxic compounds from the cell and render cells resistant to multiple drugs. Major Facilitator Superfamily (MFS) members are capable of transporting various substrates such as sugars, polyols, drugs, neurotransmitters, amino acids, peptides, and inorganic anions, although most members are substrate-specific. MFSD2B is closely related to MFSD2A, which is expressed in many tissues and is highly induced in liver and brown adipose tissue (BAT) during fasting, playing a major role in the induction of MFSD2A expression during adaptive thermogenesis. Unlike MFSD2A, MFSD2B is expressed in few tissues and at relatively low levels, and is not induced during fasting, suggesting that MFSD2B may play other roles.
Specifications
MFSD2B | |
Polyclonal | |
Unconjugated | |
MFSD2B | |
hMfsd2b; major facilitator superfamily domain containing 2B; major facilitator superfamily domain-containing protein 2B; MFSD2B; RGD1561716 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
388931 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NFX1 | |
MFSD2B | |
Recombinant protein corresponding to Human MFSD2B. Recombinant protein control fragment (Product #RP-99710). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction