Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFSD2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521050
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521050 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521050 Supplier Thermo Scientific Supplier No. PA521050
Add to Cart
Add to Cart

MFSD2B Polyclonal Antibody

MFSD2B Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MFSD2B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MFSD2B
Gene Accession No. A6NFX1
Gene Alias hMfsd2b; major facilitator superfamily domain containing 2B; major facilitator superfamily domain-containing protein 2B; MFSD2B; RGD1561716
Gene Symbols MFSD2B
Host Species Rabbit
Immunogen A 15 amino acid peptide near the center of human MFSD2B.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388931, 500624
Target Species Human, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.