Learn More
Invitrogen™ MFSD4 Polyclonal Antibody

Description
MFSD4 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | MFSD4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | MFSD4A |
| Gene Accession No. | Q8N468 |
| Gene Alias | major facilitator superfamily domain containing 4; major facilitator superfamily domain containing 4A; major facilitator superfamily domain-containing protein 4; Major facilitator superfamily domain-containing protein 4A; MFSD4; MFSD4A; UNQ3064; UNQ3064/PRO9894 |
| Gene Symbols | MFSD4A |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.