Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MFSD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598758
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598758 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598758 Supplier Invitrogen™ Supplier No. PA598758
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MFSD4 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

MFSD4 gene ontology (GO) annotation includes glucose transmembrane transporter activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFSD4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MFSD4A
Gene Accession No. Q8N468
Gene Alias major facilitator superfamily domain containing 4; major facilitator superfamily domain containing 4A; major facilitator superfamily domain-containing protein 4; Major facilitator superfamily domain-containing protein 4A; MFSD4; MFSD4A; UNQ3064; UNQ3064/PRO9894
Gene Symbols MFSD4A
Host Species Rabbit
Immunogen Recombinant Human Major facilitator superfamily domain-containing protein 4A protein (160-220AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148808
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.